missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Mouse, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Detect Mouse IgG using this Goat anti-Mouse IgG Antibody, (H+L) FITC conjugate validated for use in immunofluoresence.
Specifications
Specifications
| Antigen | Goat anti-Mouse IgG |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1.15mg/mL |
| Conjugate | FITC |
| Formulation | Lyophilized. Buffer = 0.02M Phosphate Buffer, 0.25M NaCl, pH 7.6 with 15mg/mL BSA and 0.1% Sodium azide. Reconstitute with 1mLof sterile distilled water. |
| Host Species | Goat |
| Purification Method | Affinity Purified |
| Quantity | 1 mL |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only