missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Mouse, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP124FMI
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP124FMI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP124FMI Supplier MilliporeSigma Supplier No. AP124F

Goat Polyclonal Antibody

Goat anti-Mouse IgG Antibody, (H+L) FITC Conjugated is an antibody against Mouse IgG for use in immunofluoresence.

Specifications

Antigen Goat anti-Mouse IgG
Applications Immunofluorescence
Classification Polyclonal
Concentration 2mg/mL
Conjugate FITC
Formulation Lyophilized from 0.02M phosphate buffered saline, 0.25M sodium chloride, pH 7.6, with 0.1% sodium azide and 15mg/mL bovine serum albumin. F/P Ratio:
14.5μg FITC per mg of antibody
Host Species Goat
Purification Method Affinity Purified
Quantity 2 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Protect from light
Show More Show Less

For Research Use Only