missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Mouse, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP308AMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP308AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP308AMI Supplier MilliporeSigma Supplier No. AP308A

Goat Polyclonal Antibody

This Goat anti-Mouse IgG Antibody, (H+L) Alkaline Phosphatase conjugate is validated for use in ELISA, western blotting for the detection of Mouse IgG.

Specifications

Antigen Goat anti-Mouse IgG (HL)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Alkaline Phosphatase (AP)
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C in undiluted format for 12 months. Do not Freeze.
Show More Show Less

For Research Use Only