missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Mouse, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat anti-Mouse IgG Antibody, FITC conjugate, Species Adsorbed detects level of Mouse IgG and has been published and validated for use in immunofluoresence.
Specifications
Specifications
| Antigen | Goat anti-Mouse IgG Species adsorbed |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1.3mg/mL |
| Conjugate | FITC |
| Formulation | Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, with .01% Thimerosal and .05% Sodium Azide. RECONSTITUTION: Reconstitute with 0.45mg/mL with sterile distilled water. |
| Host Species | Goat |
| Quantity | 500 μg |
| Primary or Secondary | Secondary |
| Target Species | Mouse |
| Show More |
For Research Use Only