Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Secondary Antibody, Biotin
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31805 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31805 reacts with the heavy chains of mouse IgG but not with the light chains of most mouse immunoglobulins. This antibody does not cross-react with non-immunoglobulin serum proteins or against mouse IgM. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 2mL of distilled water and centrifuge if not clear. Country of Origin: USA.
Specifications
Specifications
| Antigen | Mouse IgG Fc |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Show More |