Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Recombinant Secondary Antibody, Biotin
Goat Recombinant Monoclonal Secondary Antibody
Supplier: Invitrogen™ SA510243
Description
Specificity: reacts to the Fc region of Mouse IgG, including IgG1, IgG2a, and IgG2b. It shows very slight reaction with mouse IgG3. No cross reactivity with IgM, IgA, IgE, human IgG, rat IgG, or rabbit IgG. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Chemical Identifiers
| Mouse IgG Fc | |
| 1.0 mg/mL | |
| ELISA | |
| Biotin | |
| Goat | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Secondary | |
| Protein G |
| RMG06 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Mouse IgG. | |
| Secondary Antibody |
Specifications
| Mouse IgG Fc | |
| Recombinant Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Mouse IgG. | |
| 50 μg | |
| Secondary | |
| -20°C | |
| Liquid |
| ELISA | |
| RMG06 | |
| Biotin | |
| Goat | |
| Protein G | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |