Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, FITC
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31630 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31630 reacts with the heavy chains of mouse IgG subclasses 1, 2a, 2b, and 3, but not with light chains common to most mouse immunoglobulins. This antibody does not react against mouse IgM, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine, and rabbit serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 7.7 μg/mg; ~ 3 moles FITC per mole IgG (lot-dependent). Reconstitute with 0.75 mL of distilled water. Country of Origin: USA.
Specifications
Specifications
| Antigen | Mouse IgG Fc Cross-Adsorbed |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Show More |