missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Mouse, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP181BMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP181BMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP181BMI Supplier MilliporeSigma Supplier No. AP181B

Goat Polyclonal Antibody

Detect Mouse IgG using this Goat anti-Mouse IgG Antibody, biotin-SP conjugate, Species Adsorbed validated for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Mouse IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Biotin
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, with .01% Thimerosal and .05% Sodium Azide. RECONSTITUTION: Reconstitute with sterile distilled water.
Host Species Goat
Quantity 500 μL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only