missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Mouse, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP124AMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP124AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP124AMI Supplier MilliporeSigma Supplier No. AP124A
Only null left

Goat Polyclonal Antibody

Goat anti-Mouse IgG Antibody, Alkaline Phosphatase conjugate is an antibody against Mouse IgG for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Mouse IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Lyophilized. Buffer = 0.01M Tirs HCl, 0.25M NaCl, pH 8.0 with 15mg/mL BSA and 0.05% sodium azide. RECONSTITUTION: Reconstitute with sterile distilled water to match the volume indicated on the vial label.
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C in undiluted format for 12 months. Do not Freeze.
Show More Show Less

For Research Use Only