missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Mouse, AMCA, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat anti-Mouse IgG Antibody, AMCA conjugate, Species Adsorbed is an antibody against Mouse IgG for use in immunofluoresence.
Specifications
Specifications
| Antigen | Goat anti-Mouse IgG Species adsorbed |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | AMCA |
| Host Species | Goat |
| Quantity | 500 μg |
| Primary or Secondary | Secondary |
| Target Species | Mouse |
For Research Use Only