missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human Kappa Light Chain Secondary Antibody

Catalog No. A18855
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18855 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18855 Supplier Invitrogen™ Supplier No. A18855

Goat Polyclonal Secondary Antibody

The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human Kappa Light Chain
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration >4.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Host Species Goat
Immunogen Human Kappa Light Chain.
Purification Method Affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less