missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Human Kappa Light Chain Cross-Adsorbed Secondary Antibody
Description
Goat anti-Human Kappa Light Chain Cross-Adsorbed Secondary Antibody for ELISA, ICC/IF, Western Blot
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Human Kappa Light Chain Cross-Adsorbed |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | >4.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Human Kappa Light Chain. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mg |
| Show More |