missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) Recombinant Secondary Antibody

Catalog No. PISA510232
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PISA510232 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PISA510232 Supplier Invitrogen™ Supplier No. SA510232

Goat Recombinant Monoclonal Secondary Antibody

Specificity: reacts to Rabbit IgG. No cross reactivity with human IgG, rat IgG, or mouse IgG. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rabbit IgG (H+L)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone RMG03
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Host Species Goat
Immunogen Rabbit IgG.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage -20°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less