missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgM (Heavy chain) Recombinant Secondary Antibody, HRP

Catalog No. A54084
Encompass
Change view
Click to view available options
Quantity:
50 μL
250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A54084 250 μL
A5408450UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A54084 Supplier Invitrogen™ Supplier No. A54084

Goat Recombinant Monoclonal Secondary Antibody

This Goat anti-Human IgM (Heavy chain), Recombinant Monoclonal Secondary Antibody reacts with human IgM heavy chain with no cross reactivity to human IgG, IgA or various species immunoglobulins, including monkey IgM. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgM (Heavy chain)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 22H20L13
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with proprietary stabilizer and proprietary preservative; pH 7.4
Host Species Goat
Immunogen Purified human IgM.
Purification Method Protein G
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less