missing translation for 'onlineSavingsMsg'
Learn More

IgM Goat anti-Human, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP114PMI
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP114PMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP114PMI Supplier MilliporeSigma Supplier No. AP114P
Only null left

Goat Polyclonal Antibody

Goat anti-Human IgM Antibody, Fc5μ, HRP conjugate is an antibody against Human IgM for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Human IgM (Fc5μ)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Lyophilized. Buffer = PBS-BSA (15mg/mL) with 0.01% thimerosal. Reconstitute to 2mL with sterile distilled water.
Host Species Goat
Immunogen purified human IgM Fc5μ fragment.
Quantity 2 mL
Primary or Secondary Secondary
Target Species Human
Form Lyophilized
Show More Show Less

For Research Use Only