Learn More
Invitrogen™ Goat anti-Human IgG1 (Heavy chain) Recombinant Secondary Antibody
Description
This Goat anti-Human IgG1 (Heavy chain) Recombinant Monoclonal Antibody reacts with human IgG1 heavy chain with no cross-reactivity to human IgG2, IgG3, IgG4, IgA or IgM or various species immunoglobulins, including monkey IgG. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Specifications
Specifications
| Antigen | Human IgG1 (Heavy chain) |
| Applications | Peptide Array, ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 3H60L9 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Host Species | Goat |
| Immunogen | Purified Human IgG. |
| Purification Method | Protein G |
| Show More |