missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Human, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP112PMI
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP112PMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP112PMI Supplier MilliporeSigma Supplier No. AP112P

Goat Polyclonal Antibody

Detect Human IgG using this Goat anti-Human IgG Antibody, HRP conjugate validated for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Human IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Purified by immunoaffinity chromatography using antigen coupled to agarose beads. Lyophilized. Buffer is 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA. Contains no preservative.
Host Species Goat
Immunogen purified human IgG.
Purification Method Affinity Purified
Quantity 2 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Protect from light
Show More Show Less

For Research Use Only