missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Human, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP309PMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP309PMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP309PMI Supplier MilliporeSigma Supplier No. AP309P

Goat Polyclonal Antibody

Goat anti-Human IgG Antibody, HRP conjugate detects level of Human IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Human IgG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate HRP
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C in undiluted format for 12 months. Do not Freeze.
Show More Show Less

For Research Use Only