missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, Texas Red
Description
PA1-28827 detects Human IgG from human samples.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Human IgG (H+L) |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Texas Red |
| Formulation | PBS with 1% BSA and 0.01% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Human IgG whole molecule. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 100 μg |
| Show More |