missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, HRP
Description
This antibody reacts with human IgG and with light chains common with other human immunoglobulins as determined by IEP and ELISA techniques. This antibody may cross react with IgG from other species.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Human IgG (H+L) |
| Applications | ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | 50mM HEPES with proprietary stabilizer and 0.002% thimerosal |
| Host Species | Goat |
| Immunogen | Human IgG h+l. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mg |
| Show More |