missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, AP

Goat Polyclonal Secondary Antibody

Supplier:  Invitrogen™ PA128830

Catalog No. PA128830

Only null left

Description

Description

PA1-28830 detects Human IgG from human samples.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Chemical Identifiers

Human IgG (H+L)
4°C, do not freeze
Polyclonal
Liquid
IgG
Human
Human IgG whole molecule.
Secondary Antibody
1 mg/mL
Dot Blot, ELISA, Immunohistochemistry, Western Blot
Alkaline Phosphatase (AP)
Goat
RUO
TBS with 0.001M MgCl2, 0.0001M ZnCl2, 50% glycerol, 1% BSA and 0.1% sodium azide; pH 8.0
Secondary
Antigen affinity chromatography

Specifications

Human IgG (H+L)
Polyclonal
Alkaline Phosphatase (AP)
Goat
Antigen affinity chromatography
RUO
Human
Secondary Antibody
IgG
Dot Blot, ELISA, Immunohistochemistry, Western Blot
1 mg/mL
TBS with 0.001M MgCl2, 0.0001M ZnCl2, 50% glycerol, 1% BSA and 0.1% sodium azide; pH 8.0
Human IgG whole molecule.
100 μg
Secondary
4°C, do not freeze
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions