missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG Fc Recombinant Secondary Antibody, Alexa Fluor™ 488

Catalog No. A55747
Encompass
Change view
Click to view available options
Quantity:
50 μL
250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55747 250 μL
A5574750UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55747 Supplier Invitrogen™ Supplier No. A55747

Goat Recombinant Monoclonal Secondary Antibody

This Goat anti-Human IgG Fc Recombinant Secondary Antibody specifically detects human IgG Fc with no cross-reactivity to human IgA or IgM or various species immunoglobulins. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgG Fc
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3H8L9
Concentration 1.0 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 5mM sodium azide; pH 7.2-7.4
Host Species Goat
Immunogen Purified Human IgG.
Purification Method Protein G
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less