missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG Fab Superclonal™ Secondary Antibody

Catalog No. A5685650UL
Change view
Click to view available options
Quantity:
50 μL
250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5685650UL 50 μL
A56856 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5685650UL Supplier Invitrogen™ Supplier No. A5685650UL
Only null left

Goat Recombinant Superclonal Secondary Antibody

This Goat-anti-Human IgG Fab, Superclonal™ Recombinant Secondary Antibody detects human IgG Fab with specificity to the light chains kappa and lambda as well as the heavy chain CH1. This antibody does not detect the Fc and does not cross-react with immunoglobulins from other species. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. Superclonal™ Recombinant antibodies are a defined pool of recombinant monoclonal antibodies, manufactured by selecting and pooling individual clones to provide complementary, specific, and optimal epitope coverage to achieve enhanced performance. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgG Fab
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Goat
Immunogen Purified Human IgG.
Purification Method Protein G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less