missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Human, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ112PMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ112PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ112PMI Supplier MilliporeSigma Supplier No. AQ112P

Goat Polyclonal Antibody

Goat anti-Human IgG Antibody, F(ab′)2, HRP conjugate detects level of Human IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Human IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Affinity purified antibody. Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA and 0.01% thimerosal
Host Species Goat
Immunogen purified human IgG
Purification Method Affinity Purified
Quantity 500 μL
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C freeze-dried product until opened for up to 12 months. −20°C in undiluted aliquots for long term.
Show More Show Less

For Research Use Only