missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Human, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ112FMI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ112FMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ112FMI Supplier MilliporeSigma Supplier No. AQ112F

Goat Polyclonal Antibody

This Goat anti-Human IgG Antibody, F(ab′)2, FITC conjugate is validated for use in IF for the detection of Human IgG.

Specifications

Antigen Goat anti-Human IgG
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation Affinity purified antibody. Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, 0.01% thimerosal and 0.05% Sodium Azide
Host Species Goat
Immunogen purified human IgG
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C freeze-dried product until opened for up to 12 months. −20°C in undiluted aliquots for long term.
Show More Show Less

For Research Use Only