missing translation for 'onlineSavingsMsg'
Learn More

IgG γ chain Goat anti-Human, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP504RMI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP504RMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP504RMI Supplier MilliporeSigma Supplier No. AP504R

Goat Polyclonal Antibody

Goat anti-Human IgG γ chain Antibody, TRITC conjugate, Species Adsorbed is an antibody against Human IgG γ chain for use in ELISA and immunofluoresence.

Specifications

Antigen Goat anti-Human IgG γ chain Species adsorbed
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Rhodamine
Formulation Purified by affinity chromatography on pooled human IgG covalently linked to agarose. Liquid in 1.0mLPBS/NaN3.
Host Species Goat
Immunogen IgG γ chain. Pooled antisera from goats hyperimmunized with normal human IgG and human IgG paraproteins.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C under sterile conditions for up to 12 months. Avoid repeated freeze/thaw cycles. DO NOT FREEZE.
Show More Show Less

For Research Use Only