missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Human, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Goat anti-Human IgG Antibody, biotin conjugate is validated for use in ELISA, western blotting for the detection of Human IgG.
Specifications
Specifications
| Antigen | Goat anti-Human IgG |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | Purified by immunoaffinity chromatography using antigen coupled to agarose beads. Lyophilized. Buffer is 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.05% sodium azide. |
| Host Species | Goat |
| Immunogen | purified human IgG. |
| Purification Method | Affinity Purified |
| Quantity | 2 mL |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only