missing translation for 'onlineSavingsMsg'
Learn More

IgE Goat anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP175MI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP175MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP175MI Supplier MilliporeSigma Supplier No. AP175
Only null left

Goat Polyclonal Antibody

Goat anti-Human IgE Antibody is an antibody against Human IgE for use in ELISA, immunoprecipitation and western blotting.

Specifications

Antigen Goat anti-Human IgE
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 3.1mg/mL
Conjugate Unconjugated
Formulation Immunoglobulin, affinity purified. Liquid in 0.05M Tris-HCl (pH 7.5), 0.5M NaCl, 0.01M EDTA with 0.1% sodium azide.
Host Species Goat
Immunogen Human myeloma IgE
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 6 months.
Show More Show Less

For Research Use Only