missing translation for 'onlineSavingsMsg'
Learn More

IgA Goat anti-Human, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP120AMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP120AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP120AMI Supplier MilliporeSigma Supplier No. AP120A

Goat Polyclonal Antibody

Goat anti-Human IgA Antibody, IgG and IgM, Alkaline Phosphatase detects level of Human IgA and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Human IgA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Purified goat secondary antibody IgG in lyophilized buffer containing 0.01M Tris-HCL, 0.25M NaCl, pH 8.0 with 15mg/mL BSA and 0.05% sodium azide.
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Isotype IgG
Show More Show Less

For Research Use Only