missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgA Cross-Adsorbed Secondary Antibody DFS Item

Goat Polyclonal Secondary Antibody

Supplier:  Invitrogen™ A18789

Catalog No. A18789


Only null left

Description

Description

Goat anti-Human IgA Cross-Adsorbed Secondary Antibody for ELISA, Western Blot

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Human IgA Cross-Adsorbed
Polyclonal
Unconjugated
Goat
Affinity chromatography
RUO
Human
Secondary Antibody
IgG
ELISA, Western Blot
>4.5 mg/mL
PBS with 0.05% sodium azide; pH 7.2
Human IgA.
1 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions