missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human Ig Mix Secondary Antibody

Catalog No. H17000
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H17000 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H17000 Supplier Invitrogen™ Supplier No. H17000
Only null left

Goat Polyclonal Secondary Antibody

Goat anti-Human Ig Mix Secondary Antibody for Flow

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human Ig Mix
Applications Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Host Species Goat
Purification Method Purified
Quantity 2 mL
Regulatory Status ASR
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less