missing translation for 'onlineSavingsMsg'
Learn More

Ig κ chain Goat anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP502MI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP502MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP502MI Supplier MilliporeSigma Supplier No. AP502

Goat Polyclonal Antibody

Goat anti-Human Ig κ chain Antibody, Species Adsorbed detects level of Human Ig κ chain and has been published and validated for use in ELISA and immunofluoresence.

Specifications

Antigen Goat anti-Human Ig κ chain Species adsorbed
Applications ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified by affinity chromatography on pooled human Igs with κ light chains covalently linked to agarose. Liquid in 1.0mLof 100mM borate buffered saline, pH 8.2.
Host Species Goat
Immunogen Ig kappa chain. Pooled antisera from goats hyperimmunized with human myeloma proteins with κ light chains.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C under sterile conditions for up to 12 months
Show More Show Less

For Research Use Only