missing translation for 'onlineSavingsMsg'
Learn More

Ig κ chain Goat anti-Human, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP502BMI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP502BMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP502BMI Supplier MilliporeSigma Supplier No. AP502B

Goat Polyclonal Antibody

This Goat anti-Human Ig κ chain Antibody, biotin conjugate, Species Adsorbed is validated for use in ELISA, immunofluorescence for the detection of Human Ig κ chain.

Specifications

Antigen Goat anti-Human Ig κ chain Species adsorbed
Applications ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Purified by affinity chromatography on pooled human Igs with κ light chains covalently linked to agarose. Liquid in 2.0mLPBS/NaN3.
Host Species Goat
Immunogen pooled antisera from goats hyperimmunized with human myeloma proteins with κ light chains.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C under sterile conditions for up to 12 months
Show More Show Less

For Research Use Only