missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig λ chain Goat anti-Human, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Detect Human Ig λ chain using this Goat anti-Human Ig λ chain Antibody, biotin conjugate validated for use in ELISA.
Specifications
Specifications
| Antigen | Goat anti-Human Ig λ chain |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Formulation | Purified by affinity chromatography on pooled human Igs with lambda light chains covalently linked to agarose. Liquid in 2.0mLPBS/NaN3. |
| Host Species | Goat |
| Immunogen | pooled antisera from goats hyperimmunized with human myeloma proteins with lambda light chains. |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Show More |
For Research Use Only