missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Syrian Hamster IgG (H+L) Secondary Antibody, Texas Red

Catalog No. PIPA128821
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA128821 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA128821 Supplier Invitrogen™ Supplier No. PA128821
Only null left

Goat Polyclonal Secondary Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Anti-Hamster secondary antibodies are affinity-purified antibodies with well-characterized specificity for hamster immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Syrian Hamster IgG (H+L)
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Texas Red
Formulation PBS with 1% BSA and 0.01% sodium azide; pH 7.2
Host Species Goat
Immunogen Golden Syrian Hamster IgG whole molecule.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less