missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Guinea Pig IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 350

Catalog No. PISA510093
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PISA510093 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PISA510093 Supplier Invitrogen™ Supplier No. SA510093

Goat Polyclonal Secondary Antibody

This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, horse, human, mouse, rabbit, rat and sheep.

Anti-Guinea Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for guinea pig immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Guinea Pig IgG (H+L) Cross-Adsorbed
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate DyLight 350
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Host Species Goat
Immunogen Guinea pig IgG-heavy and light chain.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Guinea Pig
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less