missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Guinea Pig, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Goat anti-Guinea Pig IgG Antibody, biotin conjugate is validated for use in ELISA, western blotting for the detection of Guinea Pig IgG.
Specifications
Specifications
| Antigen | Goat anti-Guinea Pig IgG |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Host Species | Goat |
| Quantity | 2 mL |
| Primary or Secondary | Secondary |
| Target Species | Guinea Pig |
For Research Use Only