missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Chicken IgY (H+L) Secondary Antibody, FITC

Catalog No. PA128794
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA128794 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA128794 Supplier Invitrogen™ Supplier No. PA128794

Goat Polyclonal Secondary Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Chicken IgY (H+L)
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.01% sodium azide; pH 7.2
Host Species Goat
Immunogen Chicken IgY whole molecule.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less