missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor™ 647

Goat Polyclonal Secondary Antibody

Supplier:  Invitrogen™ A21449

Encompass

Catalog No. A21449


Only null left

Description

Description

Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 647 dye is a near-infrared-fluorescent dye with excitation ideally suited to the 647 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 647 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 647 dye molecules can be attached to proteins at high molar ratios without significant self-quenching, enabling brighter conjugates and more sensitive detection. The degree of labeling for each conjugate is typically 2-8 fluorophore molecules per IgG molecule; the exact degree of labeling is indicated on the certificate of analysis for each product lot. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. For the fluorophore-labeled antibodies a final concentration of 1-10 μg/mL should be satisfactory for most immunohistochemistry and flow cytometry applications. Product will be shipped at Room Temperature.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Chemical Identifiers

Chicken IgY (H+L)
4°C, store in dark
Polyclonal
Liquid
IgG
Chicken
Gamma Immunoglobins Heavy and Light chains.
Secondary Antibody
2 mg/mL
Immunohistochemistry, Western Blot, Immunocytochemistry
Alexa Fluor 647
Goat
RUO
PBS with 5mM sodium azide; pH 7.5
Secondary
Purified

Specifications

Chicken IgY (H+L)
Polyclonal
Alexa Fluor 647
Goat
Purified
RUO
Chicken
Secondary Antibody
IgG
Immunohistochemistry, Western Blot, Immunocytochemistry
2 mg/mL
PBS with 5mM sodium azide; pH 7.5
Gamma Immunoglobins Heavy and Light chains.
1 mg
Secondary
4°C, store in dark
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions