missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Chicken IgY (H+L) Secondary Antibody

Goat Polyclonal Secondary Antibody

Supplier:  Invitrogen™ A16056

Encompass

Catalog No. A16056


Only null left

Description

Description

The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Chemical Identifiers

Chicken IgY (H+L)
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Chicken
Chicken IgY (H+L).
Secondary Antibody
>4.5 mg/mL
ELISA
Unconjugated
Goat
RUO
PBS with 0.05% sodium azide; pH 7.2
Secondary
Affinity chromatography

Specifications

Chicken IgY (H+L)
Polyclonal
Unconjugated
Goat
Affinity chromatography
RUO
Chicken
Secondary Antibody
IgG
ELISA
>4.5 mg/mL
PBS with 0.05% sodium azide; pH 7.2
Chicken IgY (H+L).
2 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions