missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Cat Whole Serum (Forensic) Polyclonal Antibody, MP Biomedicals
Description
Host Species: Goat
Species Reactivity: Cat
Specifications
Specifications
| Antigen | Whole Serum |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Goat |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Feline |
| Product Type | Antibody |
| Form | Lyophilized |
| Show More |