missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GNPAT Monoclonal Antibody (OTI1E7)
GREENER_CHOICE

Catalog No. PIMA527400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527400 Supplier Invitrogen™ Supplier No. MA527400
Only null left

Mouse Monoclonal Antibody

GNPAT Monoclonal Antibody for Western Blot, IHC (P)

GNPAT is a key enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in GNPAT are associated with rhizomelic chondrodysplasia punctata type 2, which is characterized by rhizomelic shortening of femur and humerus, vertebral disorders, cataract, cutaneous lesions and severe mental retardation.

Specifications

Antigen GNPAT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GNPAT
Gene Accession No. O15228
Gene Alias acyl-CoA:dihydroxyacetonephosphate acyltransferase; acyl-CoA:dihydroxyacetonephosphateacyltransferase; AU019525; D1Ertd819e; DAPAT; DAP-AT; DHAPAT; DHAP-AT; Dihydroxyacetone phosphate acyltransferase; glycerone-phosphate O-acyltransferase; glyceronephosphate O-acyltransferase; GNPAT; RCDP2
Gene Symbols GNPAT
Host Species Mouse
Immunogen Full length human recombinant protein of GNPAT produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8443
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less