missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GNB2 Recombinant Rabbit Monoclonal Antibody (23GB1205)

Catalog No. MA554017
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554017 20 μL
MA554016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554017 Supplier Invitrogen™ Supplier No. MA554017
Only null left

Rabbit Recombinant Monoclonal Antibody

GNB2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. This gene contains a trinucleotide (CCG) repeat length polymorphism in its 5' UTR.

Specifications

Antigen GNB2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1205
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gnb2
Gene Accession No. P54313, P62879, P62880
Gene Alias G protein subunit beta 2; g protein subunit beta-2; G protein, beta-2 subunit; Gnb2; Gnb-2; guanine nucleotide binding protein (G protein), beta 2; guanine nucleotide binding protein (G protein), beta polypeptide 2; guanine nucleotide binding protein beta 2 subunit; guanine nucleotide binding protein, beta 2; guanine nucleotide binding protein, beta polypeptide 2; guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 2; guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-2; guanine nucleotide-binding protein, beta 2; guanine nucleotide-binding protein, beta-2 subunit; signal-transducing guanine nucleotide-binding regulatory protein beta subunit; transducin beta chain 2
Gene Symbols Gnb2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GNB2 (40-100AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14693, 2783, 81667
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less