missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNAI3, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002773B01P
Description
Sequence: MGCTLSAEDKAAVERSKMIDRNLREDGEKAAKEVKLLLLGAGESGKSTIVKQMKIIHEDGYSEDECKQYKVVVYSNTIQSIIAIIRAMGRLKIDFGEAARADDARQLFVLAGSAEEGVMTPELAGVIKRLWRDGGVQACFSRSREYQLNDSASYYLNDLDRISQSNYIPTQQDVLRTRVKTTGIVETHFTFKDLYFKMFDVGGQRSERKKWIHCFEGVTAIIFCVALSDYDLVLAEDEEMNRMHESMKLFDSICNNKWFTETSIILFLNKKDLFEEKIKRSPLTICYPEYTGSNTYEEAAAYIQCQFEDLNRRKDTKEIYTHFTCATDTKNVQFVFDAVTDVIIKNNLKECGLYSpecifications
| GNAI3 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC025285.1 | |
| GNAI3 | |
| GNAI3 (AAH25285.1, 1 a.a. ∼ 354 a.a) full-length human protein. | |
| RUO | |
| 2773 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3 | |
| 87U6/FLJ26559 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |