missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GNAI1/GNAI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA11000
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA11000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA11000 Supplier Invitrogen™ Supplier No. PA11000

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-1000 detects G protein alpha inhibitor 1/2 from human, rat, and mouse tissues and cells. PA1-1000 has been successfully used in Immunoprecipitation and Western blot procedures. By Western blot, this antibody detects an ~38 kDa protein representing G protein alpha inhibitor 1/2 from HeLa cell lysate. The PA1-1000 immunogen is a synthetic peptide corresponding to residues K(346) N N L K D C G L F(355) of human G Protein alpha Inhibitor 1/2. This sequence is completely conserved in human, rat, guinea pig, hamster, bovine, hydra, octopus, Xenopus, chicken, canine, spiny dogfish, great pond snail, starfish, and American lobster G protein alpha inhibitor 1/2. PA1-1000 immunizing peptide (Cat. # PEP-129) is available for use in neutralization and control procedures.

The G protein family of signal transducers includes the alpha, beta, gamma subunits and the effector which are most clearly distinguished by their different alpha chains. The family also includes Gs and Gi, the stimulatory and inhibitory GTP-binding regulators of adenylate cyclase; Go, a protein of unknown function that is abundantly expressed in brain; and transducin 1 and 2, proteins involved in retinal phototransduction. The G proteins possess GTPase activity and are components of a complex membrane signaling system that includes membrane-bound receptors. The system exists to modulate extracellular signals as they are transmitted into the cell.

Specifications

Antigen GNAI1/GNAI2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene GNAI2
Gene Accession No. B2RSH2, P04897, P04899, P08752, P10824, P63096
Gene Alias adenylate cyclase-inhibiting G alpha protein; AU046200; BPGTPB; C76432; G alpha inhibitory type 2 (a); G alpha inhibitory type 2 (b); G protein subunit alpha i1; G protein subunit alpha i2; Galphai2; Gi; Gi1 protein alpha subunit; Gia; Gialpha1; GIP; GNAI1; Gnai-1; GNAI1 protein; Gnai2; Gnai-2; GNAI2B; GTP-binding protein (G-alpha-i2); GTP-binding regulatory protein Gi alpha-2 chain; guanidine nucleotide binding protein, (G protein) , alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein (G protein), alpha inhibiting 2; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein, alpha inhibiting 1; guanine nucleotide binding protein, alpha inhibiting 2; guanine nucleotide binding protein, alpha inhibiting activity polypeptide 2; guanine nucleotide-binding protein G(i) subunit alpha-1; Guanine nucleotide-binding protein G(i) subunit alpha-2; guanine nucleotide-binding protein G(i), alpha-1 subunit; guanine nucleotide-binding protein G(i), alpha-2 subunit; H_LUCA15.1; H_LUCA16.1; inhibitory GTP-binding protein subunit alpha 2; p-alpha-h subunit; unnamed protein product
Gene Symbols GNAI1, GNAI2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues K(346) N N L K D C G L F(355) of human G Protein alpha Inhibitor 1/2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14677, 14678, 25686, 2770, 2771, 81664
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less