missing translation for 'onlineSavingsMsg'
Learn More

Compass Biomedical™ GMP PLUS™ Human Platelet Lysate

Advanced human platelet lysate that is safe and consistent for the rapid expansion of human mesenchymal cell cultures.

Specifications

For Use With (Application) Cell Culture, Stem Cell Culture
Purity GMP Grade
Host Species Human
Product Type Human Platelet Lysate
Content And Storage Store at -80°C upon receipt. Thaw at room temperature or in a water bath. After thawing, aliquot and store at -20°C or colder. Long term storage at -80°C is recommended. Multiple freeze/thaw cycles are not recommended.
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PLSGB0100
View Documents
Compass Biomedical™
PLSGB0100
100 mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PLSGB0250
View Documents
Compass Biomedical™
PLSGB0250
250 mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PLSGB0500
View Documents
Compass Biomedical™
PLSGB0500
500 mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PLSGB1000
View Documents
Compass Biomedical™
PLSGB1000
1000 mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Potent: Human derived product rich in growth factors, cytokine, and other proteins
  • Products are growth factor rich supplements made from large pools of transfusion grade human platelets sourced from FDA-registered/AABB accredited blood centers.
Specifications

Specifications

Cell Culture, Stem Cell Culture
Human
Store at -80°C upon receipt. Thaw at room temperature or in a water bath. After thawing, aliquot and store at -20°C or colder. Long term storage at -80°C is recommended. Multiple freeze/thaw cycles are not recommended.
GMP
GMP Grade
Human Platelet Lysate
Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'