missing translation for 'onlineSavingsMsg'
Learn More

PeproTech GM-CSF Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500p33100ug
Encompass
Change view
Click to view available options
Quantity:
100 μg
50 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P33100UG 100 μg
500P3350UG 50 μg
500P331MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 500P33100UG Supplier PeproTech Supplier No. 500P33100UG
Only null left

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GM-CSF. Anti-Human GM-CSF-specific antibody was purified by affinity chromatography employing an immobilized Human GM-CSF matrix. Sandwich ELISA: To detect Human GM-CSF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human GM-CSF (500-P33Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human GM-CSF. Western Blot: To detect hGM-CSF by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hGM-CSF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.

Specifications

Antigen GM-CSF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CSF2
Gene Accession No. P04141
Gene Alias colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim
Gene Symbols CSF2
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human GM-CSF.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1437
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less