missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GM-CSF Monoclonal Antibody (MP1-22E9), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013694
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5013694 50 μg
5013695 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5013694 Supplier Invitrogen™ Supplier No. 14733181

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

Description: The MP1-22E9 antibody reacts with mouse granulocyte/macrophage - colony stimulating factor (GM-CSF). The MP1-22E9 antibody is a neutralizing antibody. Mouse GM-CSF is a 14 kDa factor produced mainly by activated T cells and macrophages. Other cell types, such as endothelium and fibroblasts, also secrete GM-CSF in response to TNF-alpha, IL-2, IL-1, and IFN-gamma. GM-CSF stimulates growth of macrophages, granulocytes and dendritic cells. GM-CSF is found as a membrane-bound form and also as a complex associated with the extracellular matrix. Non-glycosylated GM-CSF is biologically active. Applications Reported: This MP1-22E9 antibody has been reported for use in capture of mouse GM-CSF by ELISA, intracellular staining and flow cytometric analysis, neutralization of GM-CSF bioactivity, western blotting, and immunohistochemical staining of frozen tissue sections. Applications Tested: The MP1-22E9 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse GM-CSF (Granulocyte/Macrophage-Colony Stimulating Factor, GMCSF) in combination with the biotin MP1-31G6 (13-7332) antibody for detection and recombit mouse GM-CSF (14-8331) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombit standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate.

The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.

Specifications

Antigen GM-CSF
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone MP1-22E9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CSF2
Gene Accession No. P01587
Gene Alias colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim
Gene Symbols CSF2
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12981
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.