missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GlyT2 Recombinant Rabbit Monoclonal Antibody (23GB4145)

Catalog No. MA552663
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552663 20 μL
MA552662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552663 Supplier Invitrogen™ Supplier No. MA552663
Only null left

Rabbit Recombinant Monoclonal Antibody

GlyT2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a sodium- and chloride-dependent glycine neurotransmitter transporter. This integral membrane glycoprotein is responsible for the clearance of extracellular glycine during glycine-mediated neurotransmission.

Specifications

Antigen GlyT2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4145
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC6A5
Gene Accession No. P58295, Q761V0, Q9Y345
Gene Alias glycine transporter 2; GLYT2; glyT-2; HKPX3; NET1; prestin; si:dkey-7n10.12; slc6a5; Sodium- and chloride-dependent glycine transporter 2; solute carrier family 6 (neurotransmitter transporter), member 5; solute carrier family 6 (neurotransmitter transporter, glycine), member 5; solute carrier family 6 member 5
Gene Symbols SLC6A5
Host Species Rabbit
Immunogen A synthesized peptide derived from human Glyt2 (301-336AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104245, 171148, 9152
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less