missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glypican 5 Monoclonal Antibody (K70048_12B8), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556108
Description
Sequence of this protein is as follows: EDIVKSYTQR VVGNGIKAQS GNPEVKVKGI DPVINQIIDK LKHVVQLLQG RSPKPDKWEL LQLGSGGGMV EQVSGDCDDE DGC
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. [provided by RefSeq, Jul 2008].Chemical Identifiers
| Glypican 5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| A230034F01Rik; bA93M14.1; glypican 5; glypican proteoglycan 5; glypican-5; Gpc5; Secreted glypican-5 | |
| Recombinant human GPC5 (immunogen range is 422-504). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GPC5 |
| K70048_12B8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P78333 | |
| 2262 | |
| IgG1 | |
| Protein A | |
| Antibody |
Specifications
| Glypican 5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P78333 | |
| GPC5 | |
| Recombinant human GPC5 (immunogen range is 422-504). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K70048_12B8 | |
| Unconjugated | |
| GPC5 | |
| A230034F01Rik; bA93M14.1; glypican 5; glypican proteoglycan 5; glypican-5; Gpc5; Secreted glypican-5 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2262 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |